KEY424826688 Key Organics / BIONET

6-fluoro-8-nitroquinoline

  • MDL number MFCD09907841
  • Cas Number : 343-26-0
  • Molecular Weight : 192.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433500

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4065_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart