KEY424826690 Key Organics / BIONET

8-bromo-6-methylquinoline

  • MDL number MFCD09701455
  • Cas Number : 84839-95-2
  • Molecular Weight : 222.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433502

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4067_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart