KEY424826697 Key Organics / BIONET

(2-methyl-1H-indol-5-yl)methanamine

  • MDL number MFCD05864579
  • Cas Number : 36798-25-1
  • Molecular Weight : 160.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (2-methyl-1H-indol-5-yl)methanamine
Inchi Code 1S/C10H12N2/c1-7-4-9-5-8(6-11)2-3-10(9)12-7/h2-5,12H,6,11H2,1H3
Inchi Key OZQPOLJBMYZUOL-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433509

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4083_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart