KEY424826709 Key Organics / BIONET

methyl 2-amino-3-hydroxybenzoate

  • MDL number MFCD01675233
  • Cas Number : 17672-21-8
  • Molecular Weight : 167.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name methyl 2-amino-3-hydroxybenzoate
Inchi Code 1S/C8H9NO3/c1-12-8(11)5-3-2-4-6(10)7(5)9/h2-4,10H,9H2,1H3
Inchi Key GDIJXOCHGFQHCT-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433521

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/CS-4097_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart