KEY424826712 Key Organics / BIONET

2-chloro-5-(trichloromethyl)pyridine

  • MDL number MFCD00160145
  • Cas Number : 69045-78-9
  • Molecular Weight : 230.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433524

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DK-0013_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart