KEY424826714 Key Organics / BIONET

N-(2-{[3-chloro-5-(trifluoromethyl)pyridin-2-yl]oxy}ethyl)-4-nitrobenzamide

  • MDL number MFCD28023539
  • Cas Number : 1610047-56-7
  • Molecular Weight : 389.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433526

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/DK-0022_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart