KEY424826723 Key Organics / BIONET

2-chloro-6-(trifluoromethyl)-1,3-benzoxazole

  • MDL number MFCD18250485
  • Cas Number : 1248393-79-4
  • Molecular Weight : 221.56

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point LMS
Purity 97
Storage temp. Store under N2 at 4
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433536

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411,P422
MSDS https://www.keyorganics.net/media/catalog/product/sds/DK-0710_SDS.pdf
pictograms GHS07
hstatements H302,H312,H332
Availability details for
Quantity
  • Availability
  • Add to Cart