KEY424826725 Key Organics / BIONET

4-methoxy-2,6-dimethylbenzaldehyde

  • MDL number MFCD04971199
  • Cas Number : 19447-00-8
  • Molecular Weight : 164.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

mp LMS
Iupac Name 4-methoxy-2,6-dimethylbenzaldehyde
Inchi Code 1S/C10H12O2/c1-7-4-9(12-3)5-8(2)10(7)6-11/h4-6H,1-3H3
Inchi Key ZFINNXSRPCWLHX-UHFFFAOYSA-N

Description

Melting Point LMS
Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433538

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/DK-0722_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart