KEY424826732 Key Organics / BIONET

tert-butyl 5-{4-amino-7-methyl-7H-pyrrolo[2,3-d]pyrimidin-5-yl}-2,3-dihydro-1H-indole-1-carboxylate

  • MDL number MFCD28023544
  • Cas Number : 1391053-27-2
  • Molecular Weight : 365.43

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433546

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/EK-0207_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart