KEY424826740 Key Organics / BIONET

(R)-tert-butyl[(di-tert-butylphosphanyl)methyl]methylphosphane diborane

  • MDL number MFCD12198356
  • Cas Number : 512184-96-2
  • Molecular Weight : 290.03

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433554

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0010_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart