KEY424826741 Key Organics / BIONET

(S)-tert-butyl({2-[(S)-tert-butyl(methyl)phosphanyl]ethyl})methylphosphane diborane

  • MDL number MFCD03844770
  • Cas Number : 203000-48-0
  • Molecular Weight : 261.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. Store under N2 at 4
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433555

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404,P411,P422
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0011_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart