KEY424826748 Key Organics / BIONET

(3R)-4-methoxy-3-[(naphthalen-1-yl)methyl]-4-oxobutanoic acid

  • MDL number MFCD06798981
  • Cas Number : 119807-82-8
  • Molecular Weight : 272.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433562

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0020_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart