KEY424826757 Key Organics / BIONET

(2S)-2-{[(tert-butoxy)carbonyl]amino}-3-(4-chlorophenyl)propanoic acid

  • MDL number MFCD00057808
  • Cas Number : 68090-88-0
  • Molecular Weight : 299.75

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433571

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0030_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart