KEY424826761 Key Organics / BIONET

(2R)-2-amino-3-(4-chlorophenyl)propanoic acid

  • MDL number MFCD00079675
  • Cas Number : 14091-08-8
  • Molecular Weight : 199.63

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 260
Purity 95
Storage temp. Store under N2 at -18
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433575

Safety Information

signalword Danger
pcodes P260,P262,P270,P280,P305 + P351 + P338,P402 + P404,P411,P422
hcodes H301,H312,H315,H317,H319,H332,H335
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0034_SDS.pdf
pictograms GHS06
Availability details for
Quantity
  • Availability
  • Add to Cart