KEY424826770 Key Organics / BIONET

(4Z)-2-methyl-4-[(pyridin-3-yl)methylidene]-4,5-dihydro-1,3-oxazol-5-one

  • MDL number MFCD00457206
  • Cas Number : 76315-20-3
  • Molecular Weight : 188.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Shipping Temperature RT
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433584

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/GS-0043_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart