KEY424826772 Key Organics / BIONET

1-benzyl-2,3-dihydro-1H-indole-2,3-dione

  • MDL number MFCD00187663
  • Cas Number : 1217-89-6
  • Molecular Weight : 237.258

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433586

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/GS-0045_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart