KEY424826775 Key Organics / BIONET

(S)-(+)-3,3'-Bis(3,5-bis(trifluoromethyl)phenyl)-1,1'-binaphthyl-2,2'-diyl hydrogenphosphate

  • MDL number MFCD08689863
  • Cas Number : 878111-17-2
  • Molecular Weight : 772.483

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433592

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4777_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart