KEY424826776 Key Organics / BIONET

tert-butyl (3R,4S)-3-fluoro-4-hydroxypiperidine-1-carboxylate >98% ee

  • MDL number MFCD18633012
  • Cas Number : 1174020-42-8
  • Molecular Weight : 219.256

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433593

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/SS-4802_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart