KEY424826777 Key Organics / BIONET

tert-butyl (3S,4R)-3-fluoro-4-hydroxypiperidine-1-carboxylate >98% ee

  • MDL number MFCD18632748
  • Cas Number : 1174020-40-6
  • Molecular Weight : 219.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name tert-butyl (3S,4R)-3-fluoro-4-hydroxy-1-piperidinecarboxylate
Inchi Code 1S/C10H18FNO3/c1-10(2,3)15-9(14)12-5-4-8(13)7(11)6-12/h7-8,13H,4-6H2,1-3H3/t7-,8+/m0/s1
Inchi Key XRNLYXKYODGLMI-JGVFFNPUSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/433594

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/SS-4803_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart