KEY428075735 Key Organics / BIONET

2-methyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propanamide

  • MDL number MFCD28015461
  • Cas Number : 1374263-57-6
  • Molecular Weight : 289.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form White to Off-white Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436316

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2936_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart