KEY428075736 Key Organics / BIONET

4,8-dimethyl-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1,3,6,2-dioxazaborocane

  • MDL number MFCD28015459
  • Cas Number : 1536530-95-6
  • Molecular Weight : 268.96

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436321

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2941_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart