KEY428075737 Key Organics / BIONET

(1R,2R,6S,8R)-4-(4-methoxyphenyl)-2,9,9-trimethyl-3,5-dioxa-4-boratricyclo[6.1.1.0^{2,6}]decane

  • MDL number MFCD28098208
  • Cas Number : 1258238-86-6
  • Molecular Weight : 286.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Colourless to Off-white Liquid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436322

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2949_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart