KEY428075738 Key Organics / BIONET

potassium (4-{4-[(tert-butoxy)carbonyl]piperazine-1-carbonyl}phenyl)trifluoroboranuide

  • MDL number MFCD28098210
  • Molecular Weight : 396.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436327

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2954_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart