KEY428075740 Key Organics / BIONET

2,2-dimethyl-N-[4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl]propanamide

  • MDL number MFCD09842849
  • Cas Number : 1409999-54-7
  • Molecular Weight : 303.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name N-(4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)phenyl)pivalamide
Inchi Code 1S/C17H26BNO3/c1-15(2,3)14(20)19-13-10-8-12(9-11-13)18-21-16(4,5)17(6,7)22-18/h8-11H,1-7H3,(H,19,20)
Inchi Key IGYCMBQKZPTDFG-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436329

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2956_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart