KEY428075746 Key Organics / BIONET

potassium trifluoro[4-(2-methoxyethoxy)-3-nitrophenyl]boranuide

  • MDL number MFCD28052746
  • Molecular Weight : 303.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436345

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2972_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart