KEY428075754 Key Organics / BIONET

potassium (3,4-dimethoxyphenyl)trifluoroboranuide

  • MDL number MFCD18861582
  • Cas Number : 705254-33-7
  • Molecular Weight : 244.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436311

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2928_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart