KEY428075756 Key Organics / BIONET

4-[4-(azepan-1-yl)-3-nitrophenyl]-1,2-dihydrophthalazin-1-one

  • MDL number MFCD28098206
  • Cas Number : 882000-10-4
  • Molecular Weight : 364.405

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/84468

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/6Y-0815_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart