KEY428075758 Key Organics / BIONET

potassium [4-(9H-carbazol-9-yl)phenyl]trifluoroboranuide

  • MDL number MFCD20486766
  • Cas Number : 1874177-91-9
  • Molecular Weight : 349.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name potassium [4-(9H-carbazol-9-yl)phenyl](trifluoro)borate(1-)
Inchi Code 1S/C18H12BF3N.K/c20-19(21,22)13-9-11-14(12-10-13)23-17-7-3-1-5-15(17)16-6-2-4-8-18(16)23;/h1-12H;/q-1;+1
Inchi Key NYFSUSNCESHVIN-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436331

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2958_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart