KEY428075764 Key Organics / BIONET

4,4,5,5-tetramethyl-2-(3-nitro-4-phenoxyphenyl)-1,3,2-dioxaborolane

  • MDL number MFCD22570790
  • Cas Number : 364354-15-4
  • Molecular Weight : 341.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436337

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/AS-2964_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart