KEY428075765 Key Organics / BIONET

4-bromo-2-propoxybenzonitrile

  • MDL number MFCD03944296
  • Cas Number : 588682-07-9
  • Molecular Weight : 240.10

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436338

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2965_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart