KEY428075767 Key Organics / BIONET

4-bromo-2-(3-methylbutoxy)benzonitrile

  • MDL number MFCD28098214
  • Cas Number : 1616500-68-5
  • Molecular Weight : 268.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436340

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/AS-2967_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart