KEY428075798 Key Organics / BIONET

4,6-dimethoxyquinoline-2-carboxylic acid

  • MDL number MFCD20754675
  • Cas Number : 52144-22-6
  • Molecular Weight : 233.223

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436369

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3247_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart