KEY428075805 Key Organics / BIONET

4-ethoxy-6,8-difluoroquinoline-2-carboxylic acid

  • MDL number MFCD20754667
  • Cas Number : 1351843-49-6
  • Molecular Weight : 253.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436376

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3254_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart