KEY428075810 Key Organics / BIONET

6-fluoro-4-oxo-1-propyl-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD11557456
  • Cas Number : 70032-17-6
  • Molecular Weight : 249.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-fluoro-4-oxo-1-propyl-1,4-dihydro-3-quinolinecarboxylic acid
Inchi Code 1S/C13H12FNO3/c1-2-5-15-7-10(13(17)18)12(16)9-6-8(14)3-4-11(9)15/h3-4,6-7H,2,5H2,1H3,(H,17,18)
Inchi Key AXESECUUDQNNCY-UHFFFAOYSA-N

Description

Purity 97%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436381

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3260_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart