KEY428075814 Key Organics / BIONET

methyl 3-amino-4-(2-fluorophenyl)thiophene-2-carboxylate

  • MDL number MFCD19705862
  • Cas Number : 1325307-27-4
  • Molecular Weight : 251.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436385

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3266_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart