KEY428075815 Key Organics / BIONET

methyl 3-amino-4-(2-chlorophenyl)thiophene-2-carboxylate

  • MDL number MFCD08446934
  • Cas Number : 925005-57-8
  • Molecular Weight : 267.73

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436386

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3267_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart