KEY428075820 Key Organics / BIONET

7-chloro-3-(morpholine-4-carbonyl)-1H-4lambda6,1,2-benzothiadiazine-4,4-dione

  • MDL number MFCD18804136
  • Cas Number : 1251703-64-6
  • Molecular Weight : 329.76

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436391

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3273_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart