KEY428075821 Key Organics / BIONET

3-(azepane-1-carbonyl)-1H-4lambda6,1,2-benzothiadiazine-4,4-dione

  • MDL number MFCD18804133
  • Cas Number : 1251698-96-0
  • Molecular Weight : 307.37

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436392

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3274_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart