KEY428075823 Key Organics / BIONET

ethyl 1-[(2-chloropyridin-3-yl)sulfonyl]piperidine-4-carboxylate

  • MDL number MFCD13678497
  • Cas Number : 1156669-42-9
  • Molecular Weight : 332.80

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436394

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3276_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart