KEY428075825 Key Organics / BIONET

3-(1,3-dioxo-2,3-dihydro-1H-isoindol-2-yl)-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD19705865
  • Molecular Weight : 257.205

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436396

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3279_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart