KEY428075828 Key Organics / BIONET

1-{[3-(thiophen-2-yl)-1,2,4-oxadiazol-5-yl]methyl}piperidin-3-amine hydrochloride

  • MDL number MFCD28098217
  • Cas Number : 1239757-03-9
  • Molecular Weight : 300.81

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Storage temp. RT
Physical Form Off-white to Dark Yellow Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436399

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3282_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart