KEY428075831 Key Organics / BIONET

[4-propyl-2-(1H-pyrrol-1-yl)-1,3-thiazol-5-yl]methanol

  • MDL number MFCD22383830
  • Cas Number : 1400540-67-1
  • Molecular Weight : 222.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436402

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3285_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart