KEY428075835 Key Organics / BIONET

4-[(4-methoxyphenyl)methoxy]benzaldehyde

  • MDL number MFCD03937574
  • Cas Number : 77182-73-1
  • Molecular Weight : 242.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-[(4-methoxybenzyl)oxy]benzaldehyde
Inchi Code 1S/C15H14O3/c1-17-14-6-4-13(5-7-14)11-18-15-8-2-12(10-16)3-9-15/h2-10H,11H2,1H3
Inchi Key HHHSHAZGHYCUPP-UHFFFAOYSA-N

Description

Purity 95%
Physical Form Solid
Country of Origin GB
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436406

Safety Information

signalword Warning
pcodes P264,P270,P280,P301 + P312 + P330,P302 + P352 + P312,P363,P402 + P404,P501
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3292_SDS.pdf
hcodes H302,H312,H332
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart