KEY428075836 Key Organics / BIONET

2,3-dimethyl 4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-2,3-dicarboxylate

  • MDL number MFCD20131450
  • Cas Number : 391864-58-7
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436407

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3293_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart