KEY428075838 Key Organics / BIONET

methyl 2-(hydroxymethyl)-4H,5H,6H-pyrrolo[1,2-b]pyrazole-3-carboxylate

  • MDL number MFCD26130365
  • Cas Number : 1616500-57-2
  • Molecular Weight : 196.206

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436409

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3295_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart