KEY428075839 Key Organics / BIONET

2,3-dimethyl 4H,5H,6H-pyrrolo[1,2-b]pyrazole-2,3-dicarboxylate

  • MDL number MFCD28042430
  • Cas Number : 86477-07-8
  • Molecular Weight : 224.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Storage temp. RT
Physical Form Off-white to Dark Brown Solid
Shipping Temperature RT
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436410

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/media/catalog/product/sds/BS-3296_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart