KEY428075840 Key Organics / BIONET

methyl 2-[(4-methylbenzenesulfonyl)methyl]-4H,5H,6H,7H-pyrazolo[1,5-a]pyridine-3-carboxylate

  • MDL number MFCD28053075
  • Cas Number : 1616500-59-4
  • Molecular Weight : 348.42

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436411

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3297_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart