KEY428075841 Key Organics / BIONET

4-{5-[(piperidin-4-yl)methyl]-1,2,4-oxadiazol-3-yl}pyridine

  • MDL number MFCD16874858
  • Cas Number : 1239722-79-2
  • Molecular Weight : 244.298

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436412

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/BS-3298_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart