KEY428075844 Key Organics / BIONET

2H,3H-[1,4]dioxino[2,3-c]pyridine-7-carbaldehyde

  • MDL number MFCD09907902
  • Cas Number : 443955-90-6
  • Molecular Weight : 165.148

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 114-116
Purity 97
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436415

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4102_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart