KEY428075851 Key Organics / BIONET

1-phenylprop-2-en-1-amine hydrochloride

  • MDL number MFCD16658577
  • Cas Number : 102879-25-4
  • Molecular Weight : 169.65

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.90
Country of Origin United Kingdom
Product Link https://www.keyorganics.net/bionet/catalog/product/view/id/436422

Safety Information

signalword Warning
pcodes P260,P270,P280,P402 + P404
hcodes H302,H312,H332
MSDS https://www.keyorganics.net/bionet/media/catalog/product/sds/CS-4112_SDS.pdf
pictograms GHS07
Availability details for
Quantity
  • Availability
  • Add to Cart